Digital mini thermometer with extended penetration probe
Supplier: Testo
0560.1111
TEST0560.1111EA 127 SGD
TEST0560.1111
Digital mini thermometer with extended penetration probe
Thermometers Digital Thermometers
This mini thermometer with extended penetration probe shaft is the ideal instrument for taking fast, accurate core temperatures.
  • Digital display shows the exact measured value in °C or °F
  • No reading errors
  • Battery status indicated on display
  • Easy to use and portable in size
Regardless of the field of use, it shows temperatures of up to +250 °C quickly and reliably. The 213 mm long penetration probe allows easy immersion in liquids or penetration of semi-solid substances such as food quality controls, air temperature measurements in HVAC trade.