Digital mini waterproof thermometers
Supplier: Testo
0560.1113
TEST0560.1113EA 120 SGD
TEST0560.1113
Digital mini waterproof thermometers
Thermometers Digital Thermometers
These digital mini thermometers have a measuring range from −20 to +230 °C. After the measurement, it can simply be rinsed under running water, or even cleaned in a dishwasher. Suitable for a wide range of applications, such as the food and pharmaceutical sector or in HVAC applications.
  • Waterproof with protective class IP 67
  • Battery charge level indicator
  • Maximum/minimum and 'HOLD' button
  • Protective sleeve holder
This thermometer allows to measure the air temperature and core temperature of a variety of different media such as liquid, soft and powderyMore Product Information