Precision digital thermometer, testo 112
Supplier: Testo
0560.1128
TEST0560.1128EA 700 SGD
TEST0560.1128
Precision digital thermometer, testo 112
Thermometers Digital Thermometers
The highly accurate temperature measuring instrument is ideal for official food inspections.
  • Audible alarm with adjustable limits
  • Large illuminated display
  • Display and storage of minimun and maximum values
Storage temperature: −30 to +70 °C
Delivery information: Supplied with calibration protocol and batteries but without probes.