Manometers, smart and wireless probes, testo 510i/549i
Supplier: Testo
0560.1510
TEST0560.1510EA 425 SGD
TEST0560.1510
Manometers, smart and wireless probes, testo 510i/549i
Manometers
These Smart Probes are designed to be used with smartphones and tablets. Users can read the measurements using the testo Smart Probes App installed on their smart device. The App allows you to remotely read measurements from up to six Smart Probes and easily document/report the results. It can be used for determining air velocity and volume flow when used with Pitot tubes (not included).
All measurement data is displayed as instrument readings, tables, or graphs. The measurements can be quickly saved as PDF or Excel files. The App creates custom reports which can be saved and/or shared byMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable