Testo 805i infrared thermometer, and heating, mould, and VAC Kits, operated with smartphone
Supplier: Testo
0560.1805
TEST0560.1805EA 333 SGD
TEST0560.1805
Testo 805i infrared thermometer, and heating, mould, and VAC Kits, operated with smartphone
Thermometers Digital Thermometers
The testo Smart Probes offer highest mobility and efficiency in measurement of temperature. The probes can be operated wirelessly (Bluetooth®) with the own smartphone or tablet (testo Smart Probes App available as free download).
  • Facilitates the measurement of temperature
  • Display of up to six Smart Probes at the same time
  • The App quickly visualizes measurement value changes – as a graph or a table
  • Pre-stored menus for specific applications
  • Volume flow measurement in ducts/at outlets; noncontact temperature measurement by infrared with fast image
More Product Information