Precision digital thermometer, testo 926
Supplier: Testo
0560.9261
TEST0560.9261EA 237 SGD
TEST0560.9261
Precision digital thermometer, testo 926
Thermometers Digital Thermometers
The testo 926 TE type T digital temperature meter provides you with a quick, reliable and discreet means of checking the core temperature of food in commercial kitchens, catering and other commercial and industrial environments.
  • Connectivity for an attachable and a wireless sensor
  • Wide range of optional sensors to suit your individual needs
  • User-defined thresholds – audible alarm alerts when threshold exceeded
  • Min/max values are saved automatically
  • TopSafe protective cover protects the thermometer against dirt and water ingress and impact (only included
More Product Information