Food thermometer, HACCP, testo 106
Supplier: Testo
0563.1063 0560.1063
TEST0563.1063EA 188 SGD
TEST0563.1063 TEST0560.1063
Food thermometer, HACCP, testo 106
Thermometers Digital Thermometers
This thermometer provides a fine, yet robust, measuring tip that is ideally suited for fast temperature monitoring and food inspection in hotels, large kitchens and supermarkets. The special food probe makes an almost invisible penetration hole.
  • Threshold manually set by user
  • Audible and optical alarm when limits are exceeded
  • Automatic recognition of final value: 'HOLD' function
Probe (Lר: 55×3 mm)
Delivery information: Supplied with probe protecting cover and three-volt button cell battery.
Order Now
SPECIFICATIONS
Environmentally Preferable