Waterproof digital thermometers, HACCP, testo 108 / 108-2
Supplier: Testo
0563.1080 0563.1082
TEST0563.1080EA 338 SGD
TEST0563.1080 TEST0563.1082
Waterproof digital thermometers, HACCP, testo 108 / 108-2
Thermometers Digital Thermometers
These compact thermometers are ideal for carrying out fast and reliable temperature checks during food production, processing and storage.
  • Waterproof with connected probe sensor (IP67 protected)
  • Easy to use with self-explanatory keys and large, easy-to-read display
  • Softcase (protective cover) protects the meter from impact damage and dirt
  • Hygienic: softcase and food thermometer/sensor can be easily cleaned under running water
Suitable for transportation and storage of food, in restaurants, in large-scale kitchens and in the food service industry.