Air quality monitoring kit, Testo 440
Supplier: Testo
0563.4412
TEST0563.4412EA 1920 SGD
TEST0563.4412
Air quality monitoring kit, Testo 440
Thermometers Digital Thermometers
Thanks to its glass coated laboratory probe, the Testo 440 lab kit provides accurate results even in corrosive media.
  • Structured measurement menu for long-term measurements
  • Interchangeable probe coating made of laboratory glass
  • DURAN® glass for safe use in aggressive media
  • PT100 sensor for accurate measurement results
  • Internal data storage and USB port for flexible data export
  • Can be expanded with an extensive portfolio of digital probes for versatile applications
Use the Testo 440 laboratory kit, including PT100 probe, to measure and documentMore Product Information