LED stroboscope, testo 477
Supplier: Testo
0563.4770
TEST0563.4770EA 3130 SGD
TEST0563.4770
LED stroboscope, testo 477
Tachometers
The hand-held LED stroboscope measures rotations and vibrations and facilitates measurements during operation. The stationary image enables the inspection and quality assessment of high-frequency moving parts.
  • Measurement range: Up to 300000 flashes per minute (fpm)
  • With light intensity of up to 1500 Lux
  • Trigger input and output enable connection to external systems and control by an external sensor
  • Long operating times: battery power for up to 5 h
The testo 477 LED stroboscope can also measure the rotational speed of very small and hard-to-reachMore Product Information