Probes for temperature and humidity data loggers
Supplier: Testo
0628.0023 0628.0030 0628.0006 0610.1725 0628.7503 0628.7510 0628.7516 0628.7533 0602.0493 0572.1001 0572.6172 0572.9001
TEST0628.0023EA 206 SGD
TEST0628.0023 TEST0628.0030 TEST0628.0006 TEST0610.1725 TEST0628.7503 TEST0628.7510 TEST0628.7516 TEST0628.7533 TEST0602.0493 TEST0572.1001 TEST0572.6172 TEST0572.9001
Probes for temperature and humidity data loggers
Temperature Probes
Use the humidity/temperature probes (with an appropriate measuring instrument) for the reliable measurement of relative humidity and temperature. Humidity sensor is durable and traceable to international humidity standards, such as ILAC, PTB and NIST. Thus, even after years of use you can still count on measurement results that are reliable and correct.
Order Now
SPECIFICATIONS
Environmentally Preferable