Humidity module
Supplier: Testo
0636.9736 0636.2135
TEST0636.9736EA 693 SGD
TEST0636.9736 TEST0636.2135
Humidity module
Data Loggers
Hygroscopic materials always try to form a moisture equilibrium with the surrounding air. This principle is used to measure the moisture in material by measuring moisture equilibrium.
  • Slim design (Ø 4 mm) with remote electronics
Ideal for tight measuring points, based on the moisture equilibrium measured, testo 635 automatically shows material moisture in % in the display.
Material moisture calculation is available for the following materials:
–Concrete
– High-insulation bricks
– Solid bricks
– Hard wood
– Soft wood
– Gas concrete
– Particle board
–More Product Information