Temperature indicating strips
Supplier: Testo
0646.0072 0646.0073 0646.0074 0646.0075 0646.0076 0646.0077
TEST0646.0072EA 90.3 SGD
TEST0646.0072 TEST0646.0073 TEST0646.0074 TEST0646.0075 TEST0646.0076 TEST0646.0077
Temperature indicating strips
Temperature Indicating Strips
These indicators are self-adhesive, temperature proof foils with temperature sensitive elements for temperature control and regulation. They are particularly suitable for monitoring temperature in small objects.
  • Change in colour within two seconds
  • Practical booklet with 10 indicators
  • Maximum storage temperature: 25 °C; storage in a refrigerator is recommended