pH food electrode
Supplier: Testo
0650.0245
TEST0650.0245EA 834 SGD
TEST0650.0245
pH food electrode
Electrodes pH Electrodes
Robust and dirt-insensitive pH penetration electrode without temperature sensor.
  • With hole diaphragm
  • Unbreakable plastic coating around glass electrode
  • Maintenance-free food-safe gel electrolyte
Ideal for use with Testo 230 instrument.
Delivery information: pH electrode, including fixed cable with BNC plug and wetting cap.