Mini thermometer
Supplier: Testo
0900.0530
TEST0900.0530EA 171 SGD
TEST0900.0530
Mini thermometer
Thermometers Digital Thermometers
Small and handy mini thermometer with min./max. alarm.
  • Adjustable min./max. alarm: Acoustic signal sounds when the temperature exceeds or falls below the individually adjustable limit values
  • Permanently connected probe (penetration probe with 80 cm cable)
  • Probe and cable can be stored easily on the temperature measuring instrument
  • With clip for set-up, wall mounting or attachment
The mini alarm thermometer is suitable for temperature measurement in air, in soft powdery substances and in liquids. Using the clips supplied, the mini alarm thermometer canMore Product Information