Swabs, with cotton heads
Supplier: Texwipe
STX705P
TEXWSTX705PEA 100 SGD
TEXWSTX705P
Swabs, with cotton heads
Swabs Swabs for Cleaning
Sterile cotton swabs are constructed from 100% USP grade cotton. Bonded securely to a shaft by an aqueous-based adhesive. Made to exacting and consistent tolerances using high precision automated processes.
  • Gamma irradiated pre-sterilised swabs eliminating repackaging, documentation and validation of autoclaved swabs
  • Lot number and expiration date on each package and in barcode for inventory controls
  • Certificates of compliance to confirm irradiation dose with specifications
  • Clear packaging which offers visibility of seal integrity
Packaging: Easy toMore Product Information