Cleanroom floor wipes, AlphaWipe®
Supplier: Texwipe
TX1025
TEXWTX1025EA 892 SGD
TEXWTX1025
Cleanroom floor wipes, AlphaWipe®
Wipes Cleanroom Wipes Dry Cleanroom Wipes
These mop covers are constructed from 100% continuous-filament polyester in a double-knit, no-run interlock pattern.
  • Ultra low levels of particles and extractables
  • Continuous-filament fiber prevents shedding
  • Prevents cleanroom contamination
  • Resists abrasion with rigorous use or rough surfaces
  • Excellent chemical resistance
Cleanroom laundered and packaged, AlphaWipe® is an ultraclean and highly sorbent wipe ideal for critical environments. They are designed for use with Doodleduster™ mops by 3M™ but can also be used as a work surface cover. Ideal forMore Product Information