Cleanroom wipes, pre-saturated, TechniSat®
Supplier: Texwipe
TX1041
TEXWTX1041EA 1200 SGD
TEXWTX1041
Cleanroom wipes, pre-saturated, TechniSat®
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
These pre-wetted wipes are made of a fabric blend of 55% non woven hydroentangled cellulose and 45% polyester.
  • Convenient wipes offer portion control and reduced potential of spillage in and out of the cleanroom
  • Excellent bi-directional strength and a very low extractable level
  • Ideal for cleaning environmental cleanroom surfaces during and following the production flow
  • Suitable for wiping of production equipment requiring low solvent extractables and low particle generation
This fabric combines the highly absorbent properties of a natural fibre withMore Product Information