Wipes, pre-saturated, PolySat®
Supplier: Texwipe
TX1051
TEXWTX1051EA 1070 SGD
TEXWTX1051
Wipes, pre-saturated, PolySat®
Wipes Disposable Wipes and Towels Presaturated Wipes
PolySat® is constructed from a 100% melt-blown polypropylene substrate that is pre-wetted with 70% isopropyl alcohol and packaged in a resealable FlexPack. PolySat® is convenient for cleaning sensitive surfaces and polishing equipment in controlled environments.
  • FlexPacks are a convenient alternative to wipes and spray bottles, eliminating mixing, sterilising and waste
  • Packed in SolventSafe plastic that can be wiped down prior to introduction into the cleanroom
  • Delivers a pre-measured amount of high purity alcohol to the surface: Optimises alcohol usage
More Product Information