Cleanroom wipes, AlphaSorb® HC
Supplier: Texwipe
TX2412
TEXWTX2412EA 1370 SGD
TEXWTX2412
Cleanroom wipes, AlphaSorb® HC
Wipes Cleanroom Wipes Dry Cleanroom Wipes
AlphaSorb® HC is a cleanroom laundered, 2 ply, 100% polyester wipe made from continuous-filament polyester knitted in a double-knit interlock pattern.
  • Combine high sorbency with low particle and extractable levels
  • Allow rigorous wiping with minimal release of fibres
  • Also available sterile (Please refer to website for ordering information)
Developed specifically for the pharmaceutical industry and other critical environments where high absorbency is required. Autoclavable.
Suitable for use in class 10, ISO 4 cleanrooms.
Colour: White.