Cleanroom wipes, sterile, Vertex®
Supplier: Texwipe
TX3042
TEXWTX3042EA 901 SGD
TEXWTX3042
Cleanroom wipes, sterile, Vertex®
Wipes Cleanroom Wipes Sterile Cleanroom Wipes
Vertex® wipes are the cleanest, most consistent, and most rigorously quality tested cleanroom wipes.
  • Quarterly Agar Overlay and LAL testing assures that wipes do not contribute cytotoxic leachables and are non pyrogenic
  • Meet Sterility Assurance Level of 10⁻⁶
  • Soft feel for use with delicate surfaces or applications
High sorption wipes: 100% continuous filament, polyester knit fabric wipes made in the USA from virgin stock and processed on Texwipe’s Vertex® manufacturing system, a hands-free wipe cleaning system. The wipe's unique design provides a veryMore Product Information