Cleanroom wipes, pre-saturated, sterile, Vertex®
Supplier: Texwipe
TX3042P
TEXWTX3042PEA 130 SGD
TEXWTX3042P
Cleanroom wipes, pre-saturated, sterile, Vertex®
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
These 100% continuous filament, polyester knit fabric wipes are made from virgin stock and processed on Texwipe’s Vertex® manufacturing system. The Vertex® manufacturing process cleans and converts wipes in an integrated, hands-free micro environment. The wipes are manufactured in an ISO class 3 environment with a small physical footprint which provides for a more sustainable process.
  • Sealed edge wipes
  • Special design provides a very soft feel and high sorptive capacity
  • Ideal for use with delicate surfaces or applications
  • Wipes do not contribute cytotoxic
More Product Information