Cleanroom wipes, pre-saturated, sterile, Sterile PolySat®
Supplier: Texwipe
TX3213
TEXWTX3213EA 1150 SGD
TEXWTX3213
Cleanroom wipes, pre-saturated, sterile, Sterile PolySat®
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
These wipes are constructed from a 100% melt-blown PP substrate that is pre-wetted with 70% isopropyl alcohol and 30% DIW (filtered through a 0,2 μm filter).
  • Gamma irradiated to 10⁻⁶ Sterility Assurance Level to AAMI guidelines
  • Eliminates problems associated with filtration, sterilisation and mixing of alcohol and DI water
  • Packaging allows alcohol wipe down of exterior bag before introduction into sterile suites
  • Soft, versatile wiping substrate
  • Suitable for sterile wiping of production equipment requiring low solvent extractables and low particle
More Product Information