Cleanroom wipes, pre-saturated, sterile, Sterile TechniSat®
Supplier: Texwipe
TX3214
TEXWTX3214EA 49.7 SGD
TEXWTX3214
Cleanroom wipes, pre-saturated, sterile, Sterile TechniSat®
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
These wipes constructed from a hydroentangled polyester/cellulose blend are pre-wetted with 70% isopropyl alcohol and 30% DIW (filtered through a 0,2 μm filter). Each package is gamma irradiated to 10⁻⁶ Sterility Assurance Level.
  • Binder-free, hydroentangled polyester/cellulose substrate
  • Packaging permits alcohol wipedown of exterior bag before introduction into sterile suites
  • Offers low levels of solvent extractables and particle generation
  • Ideal for wiping gloved hands
Ideal for alcohol wiping in sterile environments in pharmaceutical aseptic fillMore Product Information