Cleanroom indicator wipes, BlueWipe®
Supplier: Texwipe
TX512
TEXWTX512EA 447 SGD
TEXWTX512
Cleanroom indicator wipes, BlueWipe®
Wipes Cleanroom Wipes Dry Cleanroom Wipes
This wipe is constructed from a non woven blend of 55% cellulose and 45% polyester that offers both durability and absorbency. The blue tint indicates exposure to liquid which makes the wipe ideal for alerting the user to the exposure of liquids and/or dangerous spills.
  • Low particle count, low extractable level and solvent compatibility: Suitable for general wiping and spill control
  • Indicates exposure to liquids via blue color
  • Provides abrasion-free surface for inspection of transparent surfaces
  • Convenient C-fold packaging for quick, easy access