Spiral cleanroom notebooks, TexWrite®
Supplier: Texwipe
TX5740 TX5741 TX5742
TEXWTX5740EA 550 SGD
TEXWTX5740 TEXWTX5741 TEXWTX5742
Spiral cleanroom notebooks, TexWrite®
Notebooks
TexWrite® cleanroom spiral-bound notebook, college ruled. An economical choice for note taking and data recording in critical environments.
  • Printed with low sodium, IPA resistant inks
  • Lie flat and pages rotate freely for ease of use
Format: large (279×216 mm), medium (216×140 mm), small (127×76 mm).
Packaging: Bag-Within-A-Bag®.