Cleanroom clipboard TexWrite®
Supplier: Texwipe
TX5835
TEXWTX5835EA 661 SGD
TEXWTX5835
Cleanroom clipboard TexWrite®
Clipboards
The polypropylene cleanroom clipboard is the ideal portable writing surface for use in critical environments. The rugged clipboard is constructed from rigid polypropylene and features a druable plastic clip mechanism. Rigid back allows use while standing. This clipboard is available for use with any TexWrite® documentation material.
  • Chemical resistant, corrosion resistant, durable construction
  • Suitable for use with 8,5×11" or A4 paper
  • Securely holds 1 to 50 sheets of cleanroom paper
  • Rounded corners, no sharp edges to damage gloves or garments
  • Cleanroom packaged