Mop with handle, BetaMop™ II
Supplier: Texwipe
TX7106
TEXWTX7106EA 259 SGD
TEXWTX7106
Mop with handle, BetaMop™ II
Mops
BetaMop™ II features a tubular-knit polyester mop head for economical floor cleaning. Quick change clamp assembly.
  • High-capacity, seamless head provides high absorbency
  • Low contamination makes it ideal for floor cleaning in critical environments
  • Can be used with all wring-bucket systems