Large swabs with flexible head, CleanFoam®
Supplier: Texwipe
TX710A
TEXWTX710AEA 899 SGD
TEXWTX710A
Large swabs with flexible head, CleanFoam®
Swabs Swabs for Cleaning
Durable general-purpose swabs for use with a solvent such as isopropanol.
  • Flexible internal nylon paddle and large PU foam head with round PP handle
  • Very low in both non-volatile residue and particle generation
  • Good chemical resistance
Packaging: Cleanroom processed bag-within-a-bag.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904