Foam-covered cotton swabs, CleanTips®
Supplier: Texwipe
TX720B
TEXWTX720BEA 1250 SGD
TEXWTX720B
Foam-covered cotton swabs, CleanTips®
Swabs Swabs for Cleaning
These swabs feature a soft head with low abrasion.
  • Good chemical resistance
  • Foam head moulded around a cotton bud core on a long hardwood handle
  • Highly absorbent for general purpose use
Packaging: Bag-within-a-bag.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904