Cleaning swab, Mini Alpha®
Supplier: Texwipe
TX754B
TEXWTX754BEA 191 SGD
TEXWTX754B
Cleaning swab, Mini Alpha®
Swabs Swabs for Cleaning
Swab features 100% continuous filament polyester substrate that is heat-moulded (thermally-bonded) onto the handle, sealing exposed edges for reduced fibre release and eliminating the need for contaminating adhesives. The double layered, cleanroom-laundered, knitted Alpha polyester head is extremely clean.
  • Mini tip with a rigid paddle: Compact handle for support and control
  • Low particle generation and extractable levels
  • Excellent chemical resistance
Packaging: Cleanroom processed bag-within-a-bag.
Caution: *The product(s) on this page are outsideMore Product Information