Drying ovens, large capacity, Heratherm™ General Protocol and Advanced Protocol series
Supplier: Thermo Fisher Scientific
51029328 51029331 51029332 51029340 51029342 51029345 51029346 51029348
THBR51029328EA 8580 SGD
THBR51029328 THBR51029331 THBR51029332 THBR51029340 THBR51029342 THBR51029345 THBR51029346 THBR51029348
Drying ovens, large capacity, Heratherm™ General Protocol and Advanced Protocol series
Heratherm™ large capacity ovens were designed for larger samples or high sample volume. All models have a large, easy to view, vacuum fluorescence display and simple to use touch button operation which is controlled by a microprocessor and automatic over-temperature alarm system to protect samples. All units are available in convenient 230 V version - no special connection required.
General Protocol ovens: General Protocol ovens provide gentle heating via natural convection ideal for high capacity routine drying and heating applications. Units have corrosion-resistant stainless steel (1.40More Product Information