Autoclave tapes
Supplier: PRECISION DYNAMICS TIMEM
TSI-501-2
TIMETSI-501-2EA 21.9 SGD
TIMETSI-501-2
Autoclave tapes
Sterilisation Indicators
White adhesive crepe tape, for use in standard autoclaves.
  • Dark-coloured diagonal lines appear after 15 minutes of exposure in a steam autoclave set at +121 °C
  • Clean removal from the autoclaved object
  • Latex free
Clean removal from the autoclaved object.