Photopette® OD600 Handheld Spectrophotometer

Supplier: TIP BIOSYSTEMS
508-OD600
TIPB508-OD600EA 5400 SGD
TIPB508-OD600
Photopette® OD600 Handheld Spectrophotometer
Spectrophotometers and Photometers UV/Visible Spectrophotometers
Photopette® OD600 is an affordable handheld spectrophotometer designed to determine the cell density and monitor the growth of bacteria and yeast cultures directly in the culture medium without having to transfer the sample into a cuvette. The software comes with inbuilt calibrations and allows the user to create own custom calibrations.

  • Determine cell density and monitor cell growth
  • Quantify protein with Bradford assay
  • Direct measurements in sample - no sample transfer needed
  • Measurement precision and accuracy: ±0,003 AU
  • Measurement time: Within 2 seconds
More Product Information
New Product
Order Now

SPECIFICATIONS

Environmentally Preferable
Photometric range 0,000 - 3,000 AU
Wavelength 600 nm
Wavelength accuracy ±2 nm
Lamp source, lifetime >500000 measurements per wavelength
Languages English, german and chinese
Operating conditions Recommended: 18...25 °C at 30% RH, maximum: 5...45 °C at 80% RH (non-condensing), (25000 measurements and/or 6 days standby on a single charge), auto power save mode
Power Internal 2500 mAh lithium ion battery with microUSB charging socket
Software App running under Android, simple and intuitive to use. Factory defined applications, user defined applications generated with the 'Method Maker' function. Automatic kinetic measurements, seamless connectivity and data sharing within seconds. Geo tagging, photo, notes, date and time embedded in data files. Automatic software and firmware updates. Selftesting function.
Dimensions 202×47×42 mm
Weight ~190 g
Storage conditions −20...+50 °C (max) and up to 80% RH (non-condensing)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR