Photopette® Aqua Handheld UV-Vis Spectrophotometer - Water Testing Kit
Supplier: TIP BIOSYSTEMS
508-PA
TIPB508-PAEA 6000 SGD
TIPB508-PA
Photopette® Aqua Handheld UV-Vis Spectrophotometer - Water Testing Kit
Spectrophotometers and Photometers UV/Visible Spectrophotometers
Photopette® is a revolutionary first-of-its-kind handheld fixed wavelength spectrophotometer designed to be used in the lab or out in the field.
  • Measurement precision and accuracy: Better than ±0,003 AU
  • Measurement time: Within 2 seconds per measured wavelength
  • Photopette programmed for 525, 550 and 680nm
  • Photopette stand/holder
  • Box of non-sterile cuvetips and cuvetip ejector box (for disposed tips) - 88 total tips
Photopette® device is paired with a disposable CuveTip® and then inserted into the sample for reading absorbance. The Photopette® AquaMore Product Information