Photopette® Ultra Fast Handheld Spectrophotometer
Supplier: TIP BIOSYSTEMS
508-PB 508-PC 508-PT 508-PV
TIPB508-PBEA 9910 SGD
TIPB508-PB TIPB508-PC TIPB508-PT TIPB508-PV
Photopette® Ultra Fast Handheld Spectrophotometer
Spectrophotometers and Photometers UV/Visible Spectrophotometers
The Photopette® is the ideal instruments for quick measurements at the lab bench, in the cell culture hood or out in the plant or on the field
  • Academic teaching laboratories (life science)
  • Cell culture or tissue culture
  • DNA and Protein quantification
  • Beer, wine and spirits
  • Farming and livestock
  • Fishery industry
  • Outdoor water and chemical analysis
Photopette® Cell is a handheld spectrophotometer designed to determine the cell density of mammalian cells, e. coli, spirulina, yeast and other cells at OD600, determine lactate and ethanol concentration andMore Product Information