Spectropod® UV Microvolume Spectrophotometer
Supplier: TIP BIOSYSTEMS
508-SPUV
TIPB508-SPUVEA 9540 SGD
TIPB508-SPUV
Spectropod® UV Microvolume Spectrophotometer
Spectrophotometers and Photometers Biophotometers
The Spectropod® UV is a microvolume spectrophotometer supporting 260, 280 and 340 nm for nucleic acid (DNA, RNA, and more) and protein measurements. The instrument has a small footprint and is portable as it's powered by a lithium ion battery. It can be placed directly in the hood or whereever it is needed in the laboratory.
  • Recalibration and maintenance free
  • Bluetooth connectivity​
  • Small footprint
  • Intuitive software
  • No warm up time
  • Battery powered (Lithium ion)
  • 'Method Maker' feature for custom user calibrations
  • Data export as CSV file via e-mail or Cloud
More Product Information