Multi-position magnetic stirrers, MIX series
Supplier: 2MAG
30006_UK
TMAG30006_UKEA 7330 SGD
TMAG30006_UK
Multi-position magnetic stirrers, MIX series
Stirrers Magnetic Stirrers, Multi-position
Ultra-flat magnetic stirrers with hermetically sealed stainless steel housing. Operating temperatures up to 50 °C.
  • Maintenance- and wear-free inductive drive
  • 100% synchronised jerk-free mixing
  • SoftStart for reliable and safe acceleration of the stirring bar
  • Storage of last working status when turning off
  • Digital display for speed settings
MIX 4 MS: Four individually controlled stirring positions, synchronous speed mode for all positions
Individual and independent speed setting for each stirring drive (master-slave-technology)
On/Off function