Magnetic stirrer for cell cultures, bioMIX 1
Supplier: 2MAG
85001_UK
TMAG85001_UKEA 3540 SGD
TMAG85001_UK
Magnetic stirrer for cell cultures, bioMIX 1
Stirrers Magnetic Stirrers
Ultra-flat stirring systems for careful mixing of cell cultures. Suitable for culture flasks with spinning propeller and spinning ball systems. The hermetically closed stainless steel housing is water, dust and germ-proof. Operating temperatures up to 50 °C.
  • Jerk-free mixing
  • SoftStart for reliable and safe acceleration of the stirring bar
  • Ten step power settings 10 - 100%
  • QuickSet for setting of start and maximum speed
  • Storage of last working status when turning off
  • Digital display for speed and power settings
  • Suitable for viscous culture media