Control units
Supplier: 2MAG
90200_UK 90200
TMAG90200_UKEA 2670 SGD
TMAG90200_UK TMAG90200
Control units
Stirrers Magnetic Stirrers
Order Now
SPECIFICATIONS
Model MIXcontrol 20
Environmentally Preferable
Speed range 100 - 1600 min⁻¹
Power input 20 W
Weight 1,9 kg
W×D×H 200×155×38 mm