Cell counter, 'easy read'
Supplier: TPP
87010
TPPA87010EA 1500 SGD
TPPA87010
Cell counter, 'easy read'
Cell Counters and Flow Cytometers Manual Cell Counters
These 'easy read' measuring device are fast, easy to use, reproducible and accurate alternative to commonly used methods for determination of a cell number density. The read-out process of the cell mass in the PCV tube with this requires neither special training nor sophisticated expensive machines.
  • Accurate and reproducible cell pellet measuring
  • Solid re-usable and easy handling measuring device
  • Read-out device with magnifying glass and cross line
Optimal with the use of PCV tubes.