Cell scrapers and cell spatula
Supplier: TPP
99002
TPPA99002EA 812 SGD
TPPA99002
Cell scrapers and cell spatula
Cell Scrapers
Designed for gentle removal and harvesting of tissue cultures in all types of flasks, dishes and tubes.
  • Easier and more effective collection
  • Scraper head made from flexible PE
  • Pivoting scraper head for easy manoeuvering
  • Choice of three different blade widths
  • Small raised knobs on handle for secure grip
  • Lint-free blister packaging
  • User friendly dispenser case
The cell scraper features a pivoting head, which allows changes in blade angle while directly over the culture surface, giving accessibility to adherent cells even in difficult to reach flaskMore Product Information