Tissue Flotation Work Station™ bath, TBS®
Supplier: TRIANGLE BIOMEDICAL SCIENCES
FWS-S-220
TRIAFWS-S-220EA 11550 SGD
TRIAFWS-S-220
Tissue Flotation Work Station™ bath, TBS®
Baths Tissue Baths
Flotation Work Station™ baths is ergonomically designed to provide reliability with advanced digital microprocessor control over temperature protection for preparation of paraffin embedded tissue sections.
  • LED light array for optimum specimen viewing
  • Easier, safer and more efficient
  • Low-profile for user comfort
  • Optional slide dryer and a HISTO/Orientator™ pad for eliminating wrinkles