Mega 108 Storage Cabinet for HPLC Columns
Supplier: TRIPP
50001
TRNT50001EA 3650 SGD
TRNT50001
Mega 108 Storage Cabinet for HPLC Columns
Furniture Storage Cabinets
HPLC storage cabinet with constructed with white PVC and acrylic doors.
  • Mega organiser cabinet with seven removable, slide-out trays
  • Cabinet has a total of 108 adjustable compartments
  • Removable dividers fit most analytical HPLC columns
  • Each tray is covered with an acrylic lid that is completely removable
Ordering information: Doors with different colours are available on request.