Storage cabinet for HPLC column
Supplier: TRIPP
50004
TRNT50004EA 789 SGD
TRNT50004
Storage cabinet for HPLC column
Furniture Storage Cabinets
HPLC storage cabinet with acrylic doors.
  • White PVC and acrylic plastic construction
  • Lockable hasp
  • Holds up to 30 analytical columns
  • Mounts with two standard sized keyholes
  • The crystal clear acrylic doors make viewing a snap and columns fit easily on arms to maximise storage
Ordering information: Doors with different colours are available on request.