Precision weights, OIML, class E2
Supplier: TROEMNER INSTRUMENTS
7534-E2W
TROE7534-E2WEA 481 SGD
TROE7534-E2W
Precision weights, OIML, class E2
Weights Calibration Weights
These individual milligram weights are manufactured from the finest stainless steel. Available in flat or wire construction.
Certifications: Accredited by National Voluntary Laboratory Accreditation Programme under Lab Code 105013-0 and Statement of Accuracy.
Delivery information: Supplied in attractive, durable, high quality polycarbonate cases with a compartment for each weight within the case. There is also a compartment for a brush, forceps, and a USB flash drive which contains the electronic certificate.