Incubating/cooling orbital shaker
Supplier: TROEMNER INSTRUMENTS
980145ILC
TROE980145ILCEA 11540 SGD
TROE980145ILC
Incubating/cooling orbital shaker
Shakers Shaking Incubators
The Incubating/cooling orbital shaker for PCR heats to a higher temperature and speed than most comparable models. Designed for assays that benefit from highly precise temperature control and shaking speed, the unit accommodates up to two standard SBS PCR plates simultaneously. Calibrating to an external temperature device is possible through the integrated temperature calibration mode.
  • Triple eccentric drive handles heavy workloads, provides uniform agitation and enables continuous 24-hour operation even at high speeds
  • Precisely monitor and control
More Product Information