Dishes, evaporating, silica vitreosil
Supplier: TECHNICAL QUARTZ SOLUTIONS
73001
TSLA73001EA 186 SGD
TSLA73001
Dishes, evaporating, silica vitreosil
Trays/Dishes/Pots Evaporating Basins/Dishes
Basins with round bottom and spout made from quartz Vitreosil®.
  • Very good chemical resistance
  • Very good temperature resistance
  • Easy to clean