Crucibles, low form
Supplier: TECHNICAL QUARTZ SOLUTIONS
73060 73061 73063 73065 73067
TSLA73060EA 120 SGD
TSLA73060 TSLA73061 TSLA73063 TSLA73065 TSLA73067
Crucibles, low form
Crucibles
These crucibles are made from fused quartz, translucent, glazed.
  • Max. temperature: 1050 °C
Ordering information: Crucible and lid have to be ordered separately.
Order Now
SPECIFICATIONS
Environmentally Preferable