Crucibles, tall form
Supplier: TECHNICAL QUARTZ SOLUTIONS
73069
TSLA73069EA 160 SGD
TSLA73069
Crucibles, tall form
Crucibles
Fused quartz, translucent, glazed crucibles.
  • Max. temperature: 1050 °C
Ordering information: Crucible and lid have to be ordered separately.